Serological pipettes
About this item
High-quality transparent PS, graduated, sterile.
- Clear and easy-to-read graduation, with a vertical coloured stripe on the 1, 2, 5, 10 and 25-ml pipettes
- Increased capacity thanks to negative graduation
- Cotton insert to stop liquids from rising up into the pipetting device
- Colour code conforms to international standards to facilitate pipette selection
- Free from DNase, RNase and human DNA, non pyrogenic and non cytotoxic
Packaging: Bulk packed in plastic resealable zip sachets, individually wrapped in peelable plastic/paper, or individually packaged in peelable plastic with a practical opening and aseptic release for the pipette (batch number and use-by date printed on each item of packaging). Triple packed products are packed in three individual peel bags. The inner bag is labeled with the relevant product information, LOT number and expiry date.
16 Options Available Below
Product Details & Documents
High-quality transparent PS, graduated, sterile.
- Clear and easy-to-read graduation, with a vertical coloured stripe on the 1, 2, 5, 10 and 25-ml pipettes
- Increased capacity thanks to negative graduation
- Cotton insert to stop liquids from rising up into the pipetting device
- Colour code conforms to international standards to facilitate pipette selection
- Free from DNase, RNase and human DNA, non pyrogenic and non cytotoxic
Packaging: Bulk packed in plastic resealable zip sachets, individually wrapped in peelable plastic/paper, or individually packaged in peelable plastic with a practical opening and aseptic release for the pipette (batch number and use-by date printed on each item of packaging). Triple packed products are packed in three individual peel bags. The inner bag is labeled with the relevant product information, LOT number and expiry date.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 612-0677
- 612-3272
- 612-1011
- 612-9107
- 612-3323
- 612-1012
- 612-1019
- 612-3324
- 612-1013
- 612-1015
- GREI760107_200
- 612-3327
- 612-1017
- 612-3328
- 612-1018
- GREI760160-TRI_100
- Environmentally Preferable:
- Product Transparency and Disclosure
- Product Transparency and Disclosure
- Product Transparency and Disclosure
- Product Transparency and Disclosure
- Product Transparency and Disclosure
- Product Transparency and Disclosure
- Environmental certification:
- ACT Label (EU 43.9/UK 44.0)
- ACT Label (EU 43.9/UK 44.0)
- ACT Label (EU 39.1/UK 39.2/US 47.0)
- ACT Label (EU 39.1/UK 39.2/US 47.0)
- ACT Label (EU 38.9/UK 39.0/US 46.8)
- ACT Label (EU 38.6/UK 38.7/US 46.4)
Specifications
Product Family Options
Serological pipettes
Product Information
- CapacityDivisionSterilePackedAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.612-0677Environmentally PreferableEnvironmental certification - 1 ml0,01 ml+Individually wrapped in peelable plasticLog in for availability1 * 1.000 STSFr. 294.00Specifications:
More
Specifications
Cat. No.612-3272Environmentally PreferableEnvironmental certification - 1 ml0,01 ml+Individually wrapped in peelable plastic/paperLog in for availability1 * 1.000 STSFr. 302.00Specifications:
Product Transparency and Disclosure More
Specifications
Cat. No.612-1011Environmentally PreferableProduct Transparency and DisclosureEnvironmental certificationACT Label (EU 43.9/UK 44.0)Product Transparency and Disclosure - Specifications:
More
Specifications
Cat. No.612-9107Environmentally PreferableEnvironmental certification - 2 ml0,01 ml+Individually wrapped in peelable plasticLog in for availability1 * 1.000 STSFr. 342.00Specifications:
More
Specifications
Cat. No.612-3323Environmentally PreferableEnvironmental certification - 2 ml0,01 ml+Individually wrapped in peelable plastic/paperLog in for availability1 * 1.000 STSFr. 351.00Specifications:
Product Transparency and Disclosure More
Specifications
Cat. No.612-1012Environmentally PreferableProduct Transparency and DisclosureEnvironmental certificationACT Label (EU 43.9/UK 44.0)Product Transparency and Disclosure - Specifications:
More
Specifications
Cat. No.612-1019Environmentally PreferableEnvironmental certification - Specifications:
More
Specifications
Cat. No.612-3324Environmentally PreferableEnvironmental certification - 5 ml0,1 mlIndividually wrapped in peelable plastic/paperLog in for availability1 * 200 STSFr. 102.00Specifications:
Product Transparency and Disclosure More
Specifications
Cat. No.612-1013Environmentally PreferableProduct Transparency and DisclosureEnvironmental certificationACT Label (EU 39.1/UK 39.2/US 47.0)Product Transparency and Disclosure - 10 ml0,1 mlIndividually wrapped in peelable plastic/paperLog in for availability1 * 200 STSFr. 95.20Specifications:
Product Transparency and Disclosure More
Specifications
Cat. No.612-1015Environmentally PreferableProduct Transparency and DisclosureEnvironmental certificationACT Label (EU 39.1/UK 39.2/US 47.0)Product Transparency and Disclosure - Specifications:
More
Specifications
Cat. No.GREI760107_200Environmentally PreferableEnvironmental certification - Specifications:
More
Specifications
Cat. No.612-3327Environmentally PreferableEnvironmental certification - 25 ml0,2 mlIndividually wrapped in peelable plastic/paperLog in for availability1 * 200 STSFr. 232.00Specifications:
Product Transparency and Disclosure More
Specifications
Cat. No.612-1017Environmentally PreferableProduct Transparency and DisclosureEnvironmental certificationACT Label (EU 38.9/UK 39.0/US 46.8)Product Transparency and Disclosure - Specifications:
More
Specifications
Cat. No.612-3328Environmentally PreferableEnvironmental certification - 50 ml0,5 mlIndividually wrapped in peelable plastic/paperLog in for availability1 * 100 STSFr. 195.00Specifications:
Product Transparency and Disclosure More
Specifications
Cat. No.612-1018Environmentally PreferableProduct Transparency and DisclosureEnvironmental certificationACT Label (EU 38.6/UK 38.7/US 46.4)Product Transparency and Disclosure
Triple packed for cleanroom applications
Product Information
- CapacityDivisionSterilePackedAvailabilityPrice
- 25 ml0,2 mlIndividually wrapped (triple packed) in peelable plasticLog in for availability1 * 100 STSFr. 364.00Specifications:
More
Specifications
Cat. No.GREI760160-TRI_100Environmentally PreferableEnvironmental certification