ContactUsLinkComponent
Pipette stand, rotary, SP Bel-Art
Pipette stand, rotary, SP Bel-Art
Catalog # 612-0013
undefined
Pipette stand, rotary, SP Bel-Art
Catalog # 612-0013
Specifications
  • Ø×H:
    178×178 mm
  • Colour:
    White
  • Pk:
    1
Specifications
About this item
Pipette support stand made of white PP.
  • For 28 pipettes
  • 10 holes, 14 mm Ø
  • 18 holes, 11 mm Ø
Ø×H: 178×178 mm