ContactUsLinkComponent
Pipette jar
Pipette jar
Catalog # 612-1820
Supplier:  KARTELL
undefined
Pipette jar
Catalog # 612-1820
Supplier:  KARTELL
Specifications
  • Description:
    Pipette jar
  • Pk:
    1
Specifications
About this item
PP, with PE lid.
  • Can be used without the lid to autoclave pipettes (20 minutes at 120 °C)
  • Tight-fitting lid for protecting pipettes from dust and during transport
  • Can easily be cut down in height or used as a container for soaking contaminated pipettes
Ø×H ext/int: 82×426/410 mm