Slide dispenser
Slide dispenser, Grey, PS, 50
Catalog # 631-9110
Supplier: KARTELL
Jump to:
Product Details & Documents
This dispenser can accommodate up to 50 standard microscopy slides.
- Fingerprint-free dispensing by rotating the knurled knobs on either side of the moulded base
- Cover protecting the slides from dust
- Rubber rollers to ease out the slide, then handled by the edges
Specifications
- No. of slides:50
- Colour:Grey
- Description:Slide dispenser
- Material:PS