ContactUsLinkComponent
VWR®, Slide Holder Cabinet
VWR®, Slide Holder Cabinet
Catalog # 631-0823
Supplier:  VWR Collection
undefined
VWR®, Slide Holder Cabinet
Catalog # 631-0823
Supplier:  VWR Collection
Specifications
  • No. of slides:
    400
  • Colour:
    White
  • Description:
    Microscope slide holders
  • Material:
    Cardboard
  • Seq:
    02
  • W×D×H:
    125×75×1 mm
  • Pk:
    12
Specifications
About this item
Cardboard microscope slide holders offer instant reference and fast, fingertip retrieval.
  • Cardboard fitted with a protective plastic frame, which provides secure protection of specimens
  • Each slide holder card accepts four slides of any thickness
  • Space for identification and comments
Cards can be added, removed or changed for fast updating.