VWR® Cover Glasses, Square
Anbieter: VWR Collection
Zertifikate
Über diesen Artikel
Made of pure white Hydrolytic Class 1 glass.
- No bubbles or flaws
- Cleaned and polished
Packaging: In a plastic box with hinged lid.
Frequently Bought Together
Made of pure white Hydrolytic Class 1 glass.