ContactUsLinkComponent
Workstation and stacking system for PCR tubes and plates
Workstation and stacking system for PCR tubes and plates
Catalog # 732-4818
undefined
Workstation and stacking system for PCR tubes and plates
Catalog # 732-4818
Specifications
  • No. of Holes:
    96/48
  • Description:
    Workstation and stacking system
  • Pk:
    10
Specifications
About this item
Convenient platform for tube handling and reagent dispensing before and after sample processing.
  • Multiple workstations can be securely stacked
  • Can be stored at down to −190 °C
Holds:
96×0,2 ml V-bottom tubes
96×0,3 ml U-bottom Omnistrips
Thermo-Tube plates
Thermo-Fast® non-skirted 96-well plates
Thermo-Fast® semi-skirted 96-well plates
48×0,5 ml Thermo-Tubes