Plates, poly-D-lysine coated, Nunc™
Anbieter: Thermo Fisher Scientific
Zertifikate
Über diesen Artikel
PS, ready to use plates pre-coated with poly-D-lysine, with lid.
- Consistent growth surface from each lot ensures guaranteed performance
- High quality surface promotes cell attachment, growth and differentiation in serum-free and serum-containing media
- Stable at room temperature
W×L: 128×86 mm
Technische Daten
- Catalog No:
- 734-1177
- 734-1178
- 734-1184
- 734-1186
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Technische Daten
Frequently Bought Together