Measuring cylinders, low form, class B, borosilicate glass
Supplier: Kavalier
Certificates
About this item
Borosilicate glass, low form, class B.
- Hexagonal base
- White graduations
Certifications: ISO 4788, DIN 12 680-1
Frequently Bought Together
Borosilicate glass, low form, class B.