Measuring cylinders, tall form, class B, borosilicate glass
DURAN®, borosilicate glass, tall form, class B.
- Calibrated to contain 'ln'
- Removable HDPE base and protective collar, with spout
- Short graduation marks, blue graduations
DIN EN ISO 4788
: DIN EN ISO 4788
Frequently Bought Together