0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Respirators
Reusable Respirators
Accessories for half masks, 7700 Series
Accessories for half masks, 7700 Series
Accessories for half masks, 7700 Series
Catalog # NORHN06575008
Supplier:
Honeywell Safety
Expand image
Accessories for half masks, 7700 Series
Catalog # NORHN06575008
Supplier:
Honeywell Safety
Specifications
Connection type:
N series
Description:
Filter cartridge class P3, material: ABS - fiber glass paper, protection type: particulate
Pk:
4
Specifications
About this item
Product Details
Product Details