Order Entry
Switzerland
ContactUsLinkComponent
 
Steps, mobile
  139-9806
undefined
Steps, mobile
  139-9806

 

  • Description:
    Mobile steps
  • W×D×H:
    410×500×445 mm
  • Pk:
    1
  • Weight:
    2 kg

 

 

Robust aluminium 2×2 steps. Fitted with castors to permit movement in every direction. All edges are protected by plastic caps to prevent injuries.

  • Castors with spring bearing
  • Robust and non-slip
  • The stepladder sinks down into an immobile position as soon as weight is put on the ribbed treads, thus allowing safe use

Maximum load: 150 kg

 : TÜV and GS tested in compliance with DIN 4569