Hydrometers for low and high surface tension
Hydrometers for low and high surface tension suitable for general purpose applications.
- Calibration temperature of 20 °C
- Reading below the meniscus
- For low and high surface tension
- Catalog No:
- 614-0040
- 614-0033
- 614-0038
- 614-0025
- 614-0031
- 614-0034
- 614-0028
- 614-0041
- 614-0026
- 614-0023
- 614-0009
- 614-0036
- 614-0029
- 614-0035
- 614-0039
- 614-0032
- 614-0027
- 614-0030
- Length:
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
- 300 mm
Frequently Bought Together