VWR® Safety-Blue™, Thermo-Hydrometers
Supplier: VWR Collection
Certificates
About this item
These safety sugar scale combined form thermo-hydrometers can determine the specific gravity, Brix and alcohol by volume.
- Lead free ballast and glass with mercury-free blue spirit organic liquid fill
- Individually serialised, safe and easy to clean up in the event of a breakage
- Temperature standardisation of 20 °C with thermometer
Designed to measure sucrose in the food and beverage and other industries.
Certifications: ASTM
Delivery: Includes a multi-language statement of accuracy indicating traceability to NIST.
Caution: Combined form hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures. Enclosed thermometers should be used as a general reference only. For temperature critical applications, a calibrated standard thermometer is recommended.
Frequently Bought Together