Spirit lamp, DURAN®
Soda-lime glass with ground glass joint cap. The spirit lamp is an alternative to the gas-powered Bunsen burner and is also suitable for heating cooking glasses and the materials they contain.
Capacity: 100 ml.
Ø×H: 75×103 mm.
: Supplied without socket and wick.
Frequently Bought Together