Order Entry
Switzerland
ContactUsLinkComponent
 
Burner head, butterfly design
  451-0089
undefined
Burner head, butterfly design
  451-0089

 

  • Description:
    Burner head
  • Head Ø:
    17 mm
  • Pk:
    1

 

 

Nickel plated brass.

For burner head Ø 17 mm
Width of opening: 50 mm