Orders LinkContactUsLinkComponent
Goggles, V-Maxx
Goggles, V-Maxx
Supplier:   HONEYWELL SAFETY
Goggles, V-Maxx
Supplier:  HONEYWELL SAFETY
Certificates
Goggles, V-Maxx
7 Options Available Below
Product Details & Documents
Sporty design goggles with elastic strap.
  • 180° field of vision for distortion-free peripheral vision
  • Rotating strap attachments for easier use with hardhats
  • Indirect ventilation complies with splash-resistance test
Can be worn over prescription eyewear and some half mask respirators.
Weight: 93 g.
Certifications: EN 166, EN 169, EN 170, 1 F, C, 3, 4, 9
Product Family Options
Product Information
  • Type
    Lenses
    Colour
    Availability
    Price
  • Elastic strap; anti-UV (2-1,2), anti-scratch, anti-fog
    Clear PC
    Transparent
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 17.00
    SFr. 17.00
    1 * 1 ST
       SFr. 17.00
  • Elastic strap; ventilated; anti-UV (2-1,2), anti-scratch, anti-fog
    Clear PC
    Transparent
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 18.00
    SFr. 18.00
    1 * 1 ST
       SFr. 18.00
  • Neoprene strap; ventilated; anti-UV (2-1,2), anti-scratch, anti-fog
    Clear PC
    Transparent
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 17.00
    SFr. 17.00
    1 * 1 ST
       SFr. 17.00
  • Elastic strap; no ventilation; anti-UV (2-1,2), anti-scratch, anti-fog
    Clear PC
    Transparent
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 15.50
    SFr. 15.50
    1 * 1 ST
       SFr. 15.50
  • Elastic strap; no ventilation; anti-UV (2-1,2), anti-scratch, anti-fog
    Clear PC
    Transparent
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 20.60
    SFr. 20.60
    1 * 1 ST
       SFr. 20.60
  • Neoprene strap; ventilated; anti-UV (2-1,2), anti-scratch, anti-fog
    Clear acetate
    Transparent
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 22.80
    SFr. 22.80
    1 * 1 ST
       SFr. 22.80
  • Welding goggles; anti-UV (5)
    Green (shade 5) PC
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 24.40
    SFr. 24.40
    1 * 1 ST
       SFr. 24.40