Knitted gloves, HyFlex® 11-600 / 11-601 / 11-605
Supplier: Ansell
Certificates
About this item
The stretch knitted nylon liner, together with the formulation of the polyurethane coating, offers flexibility and elasticity.
- Optimal tactility and dexterity: Ultra-thin with superior tear resistance
- Polyurethane coating gives a good grip in dry conditions
- Taint and lint-free
These industrial gloves are ideal for applications where precision and/or clean handling are required and for light and delicate applications, such as assembly of small pieces or for use in cleanrooms.
Certifications: 11-600/11-601: EN 388:2016 (3121A), Cat. II
11-605: Cat. II.
11-605: Cat. II.
Specifications
- Catalog No:
- 112-0199
- 112-5073
- 112-0198
- 112-0200
- ANSE11-600/5
- 112-0203
- 112-0202
- 112-0201
- 112-0204
- ANSE11-601/5
- 112-0196
- 112-0197
- 112-4246
- 112-0195
- 112-5076
- 112-5074
- 112-5075
- 112-5077
- Model:
- 11-600
- 11-601
- 11-600
- 11-605
- 11-600
- 11-605
- 11-605
- 11-605
- 11-605
- 11-601
- 11-600
- 11-600
- 11-600
- 11-600
- 11-601
- 11-601
- 11-601
- 11-601
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Gauge:
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- EN norm:
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)
- EN 388:2016 (3121A)