ContactUsLinkComponent
Chemical resistant gloves, AlphaTec® 15-554
 :  Ansell
undefined
Chemical resistant gloves, AlphaTec® 15-554
 :  Ansell
  • Catalog No:
  • 112-0269
  • 112-2180
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Liner material:
  • Cotton
  • Cotton
  • Finish:
  • Smooth
  • Smooth
  • Cuff style:
  • Gauntlet
  • Gauntlet
  • EN norm:
  • EN ISO 374-1:2016 Type A (BCDFIJ), EN ISO 374-5:2016, EN 388:2016 (3121A)
  • EN ISO 374-1:2016 Type A (BCDFIJ), EN ISO 374-5:2016, EN 388:2016 (3121A)
  • Anti-static:
  • +
  • +
Frequently Bought Together