Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Finger cots, nitrile, LNR

Finger cots, nitrile, LNR

L
Catalog # 112-1154
Supplier:   Honeywell Safety
Finger cots, nitrile, LNR
Catalog # 112-1154
Supplier:  Honeywell Safety
Finger cots, nitrile, LNR
Finger cots, nitrile, LNR

L

Price
SFr. 121.00
Log in for availability
1 * 720 ST
Product Details & Documents

Finger cots made of comfortable and stretchy nitrile with anti-static properties. Ideal for cleanroom applications, pharmaceutical industries and electronics assembly.

  • Powder-free, smooth finish
  • Low particle count; eliminates damaging static discharge in many applications
  • Very low extractables mean less contamination from reactive ions
  • Excellent tactility and uniform thickness

Length: 67 mm

Thickness: 0,076 mm

Colour: Translucent

AQL 2,5

Certifications: AQL 2,5; Cat. I

Specifications
  • Size:
    L
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.