Examination gloves, latex, Manufix® and Manufix® Sensitive
: B. Braun
Ambidextrous examination gloves with highly elastic natural latex beaded cuff. Suitable for demanding medical applications as well as for food applications.
- Non sterile
- Manufix® gloves are powdered
- Manufix® Sensitive gloves are not powdered but have a synthetic inner coating that makes them easy to pull on
- Thiuram-free; non-allergenic
- Micro-roughened surface and textured fingertips provide secure grip
: EN 455; AQL 1,5
- Catalog No:
- 112-2117
- 112-2115
- 112-2116
- 112-3424
- 112-3423
- Model:
- Manufix® Sensitive
- Manufix® Sensitive
- Manufix® Sensitive
- Manufix® Sensitive
- Manufix® Sensitive
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
Frequently Bought Together