Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
Separating funnels, Gilson
Separating funnels, Gilson
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
Documents
Product Family Options
With solid glass key
  • Capacity
    Socket size
  • 50 ml
    19/26 NS
    1 * 1 ST
    1 * 1 ST - SFr. 125.00
    SFr. 125.00
    1 * 1 ST
       SFr. 125.00
  • 100 ml
    19/26 NS
    1 * 1 ST
    1 * 1 ST - SFr. 129.00
    SFr. 129.00
    1 * 1 ST
       SFr. 129.00
  • 250 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - SFr. 137.00
    SFr. 137.00
    1 * 1 ST
       SFr. 137.00
  • 500 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - SFr. 163.00
    SFr. 163.00
    1 * 1 ST
       SFr. 163.00
  • 1000 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - SFr. 218.00
    SFr. 218.00
    1 * 1 ST
       SFr. 218.00
  • 2000 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - SFr. 337.00
    SFr. 337.00
    1 * 1 ST
       SFr. 337.00
With PTFE key
  • Capacity
    Socket size
  • 50 ml
    19/26 NS
    1 * 1 ST
    1 * 1 ST - SFr. 144.00
    SFr. 144.00
    1 * 1 ST
       SFr. 144.00
  • 100 ml
    19/26 NS
    1 * 1 ST
    1 * 1 ST - SFr. 154.00
    SFr. 154.00
    1 * 1 ST
       SFr. 154.00
  • 250 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - SFr. 161.00
    SFr. 161.00
    1 * 1 ST
       SFr. 161.00
  • 500 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - SFr. 206.00
    SFr. 206.00
    1 * 1 ST
       SFr. 206.00
  • 1000 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - SFr. 261.00
    SFr. 261.00
    1 * 1 ST
       SFr. 261.00
  • 2000 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - SFr. 371.00
    SFr. 371.00
    1 * 1 ST
       SFr. 371.00