Orders LinkContactUsLinkComponent
Filtration funnels, Quickfit®
Filtration funnels, Quickfit®
Supplier:   SciLabware
Filtration funnels, Quickfit®
Supplier:  SciLabware
Certificates
Filtration funnels, Quickfit®
Filtration funnels, Quickfit®
2 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc.
Cone connection.
Product Family Options
Product Information
  • Porosity
    Cone size
    Plate Ø
    Availability
    Price
  • P40
    19/26 NS
    20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 142.00
    SFr. 142.00
    1 * 1 ST
       SFr. 142.00
  • P40
    19/26 NS
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 144.00
    SFr. 144.00
    1 * 1 ST
       SFr. 144.00