DURAN® Funnels, Powder
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Good chemical and physical properties
- Use in applications that require good thermal resistance
- Quick and easy to clean
With smooth internal walls. For transferring powdery materials. Can also be used for liquids. Funnel lip is ground and fire-polished.
Frequently Bought Together