Funnels, industrial, Azlon®
Supplier: SciLabware
Certificates
About this item
HDPE, translucent.
- Good chemical and temperature resistance
- Robust, rigid design
- Feature straight sided upper position to prevent spillage
With smooth internal walls. Multipurpose funnel for a wide variety of uses. Suspension eyelet for easy storage.
Frequently Bought Together