Orders LinkContactUsLinkComponent
Funnels with short stem
Funnels with short stem
Supplier:   SciLabware
Funnels with short stem
Supplier:  SciLabware
Certificates
Funnels with short stem
Funnels with short stem
8 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls.
Product Family Options
Product Information
  • Stem length
    Stem O-Ø
    For filter Ø
    Funnel O-Ø
    Availability
    Price
  • 100 mm
    10 mm
    150 - 185 mm
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 29.90
    SFr. 29.90
    1 * 1 ST
       SFr. 29.90
  • 125 mm
    12 mm
    185 - 240 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 51.60
    SFr. 51.60
    1 * 1 ST
       SFr. 51.60
  • 150 mm
    16 mm
    240 - 270 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 53.10
    SFr. 53.10
    1 * 1 ST
       SFr. 53.10
  • 175 mm
    24 mm
    320 - 385 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 129.00
    SFr. 129.00
    1 * 1 ST
       SFr. 129.00
  • 35 mm
    8 mm
    45 - 55 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 176.00
    SFr. 176.00
    1 * 10 ST
       SFr. 176.00
  • 80 mm
    8 mm
    90 - 150 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 298.00
    SFr. 298.00
    1 * 10 ST
       SFr. 298.00
  • 45 mm
    8 mm
    55 - 70 mm
    45 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 178.00
    SFr. 178.00
    1 * 10 ST
       SFr. 178.00
  • 55 mm
    8 mm
    70 - 90 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 238.00
    SFr. 238.00
    1 * 10 ST
       SFr. 238.00