Orders LinkContactUsLinkComponent
Funnels with short stem
Supplier:   VITLAB
Funnels with short stem
Supplier:  VITLAB
Certificates
Funnels with short stem
Funnels with short stem
8 Options Available Below
Product Details & Documents
PP, translucent.
  • Very good chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
With smooth interior surfaces.
Product Family Options
Product Information
  • Length
    Stem length
    Stem O-Ø
    Funnel O-Ø
    Availability
    Price
  • 45 mm
    25 mm
    1,5 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1.20
    SFr. 1.20
    1 * 1 ST
       SFr. 1.20
  • 47 mm
    25 mm
    5 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - SFr. 9.60
    SFr. 9.60
    1 * 6 ST
       SFr. 9.60
  • 65 mm
    35 mm
    5 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - SFr. 10.60
    SFr. 10.60
    1 * 6 ST
       SFr. 10.60
  • 85 mm
    43 mm
    9 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1.70
    SFr. 1.70
    1 * 1 ST
       SFr. 1.70
  • 110 mm
    55 mm
    10 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2.00
    SFr. 2.00
    1 * 1 ST
       SFr. 2.00
  • 155 mm
    77 mm
    13 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.30
    SFr. 3.30
    1 * 1 ST
       SFr. 3.30
  • 180 mm
    90 mm
    14 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 4.60
    SFr. 4.60
    1 * 1 ST
       SFr. 4.60
  • 220 mm
    95 mm
    17 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 6.30
    SFr. 6.30
    1 * 1 ST
       SFr. 6.30