Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Supplier:   BOCHEM
Forceps, precision
Supplier:  BOCHEM
Certificates
Forceps, precision
Forceps, precision
2 Options Available Below
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. Extremely precise forceps designed for a wide variety of uses in the laboratory, production etc.
  • Extremely sharp tips without grooves
  • Reverse action; closed normally, squeeze to open
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Straight, extra sharp
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 12.50
    SFr. 12.50
    1 * 1 ST
       SFr. 12.50
  • Curved, extra sharp
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 12.50
    SFr. 12.50
    1 * 1 ST
       SFr. 12.50