Rotary evaporating flasks
Supplier: Lenz Laborglas
Jump to:
About this item
DURAN®, borosilicate glass, clear or plastic-coated.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.
18 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear or plastic-coated.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.
Product Family Options
Uncoated
Product Information
- CapacitySocket sizeAvailabilityPrice
Coated
Product Information
- CapacitySocket sizeAvailabilityPrice