ContactUsLinkComponent
Flasks, iodine with collar and iodine stopper
Flasks, iodine with collar and iodine stopper
Catalog # 201-1162
Flasks, iodine with collar and iodine stopper
Catalog # 201-1162
Specifications
  • Ext.Ø:
    85 mm
  • Socket size:
    29/32 NS
  • Capacity:
    250 ml
  • Pk:
    1
Specifications
About this item
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.