ContactUsLinkComponent
Cell culture bottles, Breed-Demeter, square, DURAN®
Cell culture bottles, Breed-Demeter, square, DURAN®
Catalog # 391-0690
Supplier:  DWK Life Sciences
undefined
Cell culture bottles, Breed-Demeter, square, DURAN®
Catalog # 391-0690
Supplier:  DWK Life Sciences
Specifications
  • Capacity:
    180 ml
  • W×D×H:
    48×48×148 mm
  • Pk:
    1
Specifications
About this item
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For cell and tissue culture.