Fernbach culture flasks, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With beaded rim. Providing the large liquid surface area necessary for growing microorganisms in liquid culture media. Provides for faster growth through improved ventilation.
Frequently Bought Together