Culture bottles and culture flasks, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Culture bottles and culture flasks (erlenmeyer shape) with straight rim, suitable for either kapsenberg caps or metal caps (available separately).
Frequently Bought Together