Culture media bottles, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With beaded rim or straight neck for metal caps. Marked with capacity and with a white labelling area.
Frequently Bought Together