Orders LinkContactUsLinkComponent
Erlenmeyer flasks, narrow neck, with standard socket
Erlenmeyer flasks, narrow neck, with standard socket
Supplier:   KARTELL
Erlenmeyer flasks, narrow neck, with standard socket
Supplier:  KARTELL
Certificates
Erlenmeyer flasks, narrow neck, with standard socket
Erlenmeyer flasks, narrow neck, with standard socket
6 Options Available Below
Product Details & Documents
PP, transparent.
  • Very good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning
With moulded graduations. Autoclavable. Supplied without stoppers.
Product Family Options
Product Information
  • Capacity
    Graduation Interval
    Socket size
    Availability
    Price
  • 50 ml
    10 ml
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 5.30
    SFr. 5.30
    1 * 1 ST
       SFr. 5.30
  • 125 ml
    25 ml
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 6.80
    SFr. 6.80
    1 * 1 ST
       SFr. 6.80
  • 250 ml
    25 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 7.90
    SFr. 7.90
    1 * 1 ST
       SFr. 7.90
  • 500 ml
    50 ml
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 12.70
    SFr. 12.70
    1 * 1 ST
       SFr. 12.70
  • 1000 ml
    50 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 21.70
    SFr. 21.70
    1 * 1 ST
       SFr. 21.70
  • 2000 ml
    100 ml
    35/34 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 30.80
    SFr. 30.80
    1 * 1 ST
       SFr. 30.80