Orders LinkContactUsLinkComponent
Erlenmeyer flasks with screw caps
Erlenmeyer flasks with screw caps
Supplier:   Lenz Laborglas
Erlenmeyer flasks with screw caps
Supplier:  Lenz Laborglas
Certificates
Erlenmeyer flasks with screw caps
Erlenmeyer flasks with screw caps
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with DIN thread and red PBT closure.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and a white labelling area. Screw cap with PTFE coated seal.
Documents
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Thread
    Availability
    Price
  • 100 ml
    109 mm
    64 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 58.90
    SFr. 58.90
    1 * 1 ST
       SFr. 58.90
  • 250 ml
    145 mm
    85 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 60.50
    SFr. 60.50
    1 * 1 ST
       SFr. 60.50
  • 500 ml
    180 mm
    105 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 63.80
    SFr. 63.80
    1 * 1 ST
       SFr. 63.80
  • 1000 ml
    225 mm
    131 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 95.90
    SFr. 95.90
    1 * 1 ST
       SFr. 95.90