Order Entry
Switzerland
ContactUsLinkComponent
 
In-Line filters, Polydisc™ SPF, Whatman™
  516-1107
undefined
In-Line filters, Polydisc™ SPF, Whatman™
  516-1107

 

  • Ø:
    50 mm
  • Filtration area:
    20,4 cm²
  • Pore size:
    1,0 µm
  • Sterile:
    -
  • Seq:
    05
  • Environmentally Preferable:
  • Pk:
    10

 

 

These disc filters contain a glass microfiber (GMF) and polyethersulfone (PES) stack designed to purify and sterilise serum and other hard to filter solutions. GMF prefilter captures large particles and cell debris while PES stack removes remaining particles and bacteria larger than 1 µm.

  • High-throughput, in-line prefilters for use upstream of serum filters
  • GMF prefilter captures large particles and cell debris while PES stack removes remaining particles and bacteria larger than 1 µm
  • Designed to extend the life of downstream serum filters
  • Effective for microbiology and tissue culture, immunoassays, virology, and diagnostic controls
  • 6 to 10 mm Int.Ø hose connection

Filtering serum requires removing proteins, lipids, salts, and other cell debris. This range of particulate matter is effectively handled with multi-layer prefilters to facilitate downstream work and to avoid clogging later serum filters.

Polydisc SPF stacks a high-flow, hydrophilic PES membrane with a high particle-loading GMF filter to clean out particulates from serum and reduce stress on the final-stage serum filters.

The 1 µm device features a PP prefilter for use with heavily contaminated samples.

Integrity testable