ContactUsLinkComponent
Filter capsules with Versapor® membrane
Filter capsules with Versapor® membrane
Certificates
undefined
Filter capsules with Versapor® membrane
Certificates
Specifications
  • Catalog No:
  • 514-4093
  • 516-7521
  • 516-7523
  • Type:
  • Serum capsule
  • Mini-capsule
  • Versaflow™
  • Water flow rate:
  • 11,42 l/min (0,5 bar)
  • 11,42 l/min (0,5 bar)
  • Max. temperature:
  • 88 °C (2,1 bar)
  • 88 °C (2,1 bar)
  • Max. pressure:
  • 3,4 bar
  • 3,4 bar