Vacuum filter holders, stainless steel
For colony counting and particle collection, specially designed for applications in which the particles or microorganisms retained on the membrane filter surface are of interest. The stainless steel frit filter support ensures a uniform distribution of the residues.
Autoclavable at 134 °C.
Outlet Ø: 10 mm
Composition: with cover, clamp and tap in stainless steel.
Frequently Bought Together