Order Entry
Switzerland
ContactUsLinkComponent
Pressure filtration holder, stainless steel, 200 ml
Pressure filtration holder, stainless steel, 200 ml
Catalog # 512-3420
Supplier:  Sartorius
undefined
Pressure filtration holder, stainless steel, 200 ml
Catalog # 512-3420
Supplier:  Sartorius

Specifications

  • For Ø:
    47 mm
  • Capacity:
    200 ml
  • Material:
    Stainless steel
  • Seq:
    01
  • Max. operating pressure:
    10 bar
  • Filtration area:
    13 cm²
  • Weight:
    960 g
  • Pk:
    1

Specifications

About this item

For the filtration of small volumes, the holder is connected directly to a pressure source. For the filtration of up to 5 litre volumes of relatively easily filtrable liquids (e.g. buffer solutions, solutions for cell counters and tissue culture solutions), it is used in combination with a pressure tank.

  • Large filling opening to simplify addition of sample
  • Can be attached to a tripod and clamped down
  • With hose nipples

For the filtration of small volumes the holder is connected directly to a pressure source. For the filtration of up to 5 litre volumes of relatively easily filtrable liquids (e.g. buffer solutions, solutions for cell counters and tissue culture solutions), it is used in combination with a pressure tank. Large filling opening to simplify addition of sample. Can be attached to a tripod and clamped down.

With hose nipples

Autoclavable at max. +134 °C

Max. pressure: 10 bar