Orders LinkContactUsLinkComponent
Extraction thimbles, Munktell
Extraction thimbles, Munktell
Supplier:   Ahlstrom
Extraction thimbles, Munktell
Supplier:  Ahlstrom
Certificates
Extraction thimbles, Munktell
Extraction thimbles, Munktell
43 Options Available Below
Product Details & Documents
Grade ET/401
Particularly suitable for the extraction of organic combinations out of reaction mixtures, food, natural substances, lacquer and binder analysis in paint colours, dust and tar determination in gases.
Max. Temperature: 120 °C.
Grade MK 360
A special development for the analysis of air pollution.
Max. Temperature: 900 °C
Grade MG160
Filter thimbles of high purity particularly recommended for aggressive solvents. For analytical registration of environmental pollution.
Max. Temperature: 500 °C
T* indicates is used for Tecator machines
Product Family Options
Grade 401, cellulose
Product Information
  • Int.Ø×H
    Availability
    Price
  • 10×50 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 132.00 SFr.
    132.00 SFr.
    1 * 25 ST
       132.00 SFr.
  • 22×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 104.00 SFr.
    104.00 SFr.
    1 * 25 ST
       104.00 SFr.
  • 25×100 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 110.00 SFr.
    110.00 SFr.
    1 * 25 ST
       110.00 SFr.
  • 26×60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 109.00 SFr.
    109.00 SFr.
    1 * 25 ST
       109.00 SFr.
  • 26×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 100.00 SFr.
    100.00 SFr.
    1 * 25 ST
       100.00 SFr.
  • 30×77 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 130.00 SFr.
    130.00 SFr.
    1 * 25 ST
       130.00 SFr.
  • 30×95 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 118.00 SFr.
    118.00 SFr.
    1 * 25 ST
       118.00 SFr.
  • 35×150 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 157.00 SFr.
    157.00 SFr.
    1 * 25 ST
       157.00 SFr.
  • 33×205 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 204.00 SFr.
    204.00 SFr.
    1 * 25 ST
       204.00 SFr.
  • 43×123 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 194.00 SFr.
    194.00 SFr.
    1 * 25 ST
       194.00 SFr.
Grade MK 360, quartz microfibre
Product Information
  • Int.Ø×H
    Availability
    Price
  • 19×90 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 539.00 SFr.
    539.00 SFr.
    1 * 25 ST
       539.00 SFr.
  • 22×62 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 557.00 SFr.
    557.00 SFr.
    1 * 25 ST
       557.00 SFr.
  • 22×100 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 576.00 SFr.
    576.00 SFr.
    1 * 25 ST
       576.00 SFr.
  • 22×68 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 576.00 SFr.
    576.00 SFr.
    1 * 25 ST
       576.00 SFr.
  • 25×90 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 539.00 SFr.
    539.00 SFr.
    1 * 25 ST
       539.00 SFr.
  • 25×100 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 547.00 SFr.
    547.00 SFr.
    1 * 25 ST
       547.00 SFr.
  • 26×60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 539.00 SFr.
    539.00 SFr.
    1 * 25 ST
       539.00 SFr.
  • 30×77 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 574.00 SFr.
    574.00 SFr.
    1 * 25 ST
       574.00 SFr.
  • 30×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 574.00 SFr.
    574.00 SFr.
    1 * 25 ST
       574.00 SFr.
  • 30×100 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 586.00 SFr.
    586.00 SFr.
    1 * 25 ST
       586.00 SFr.
  • 30×110 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 677.00 SFr.
    677.00 SFr.
    1 * 25 ST
       677.00 SFr.
  • 34×150 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 622.00 SFr.
    622.00 SFr.
    1 * 25 ST
       622.00 SFr.
  • 35×150 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 574.00 SFr.
    574.00 SFr.
    1 * 25 ST
       574.00 SFr.
  • 43×123 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 625.00 SFr.
    625.00 SFr.
    1 * 25 ST
       625.00 SFr.
Grade MG160, glass microfibre
Product Information
  • Int.Ø×H
    Availability
    Price
  • 10×50 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 185.00 SFr.
    185.00 SFr.
    1 * 25 ST
       185.00 SFr.
  • 19×90 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 195.00 SFr.
    195.00 SFr.
    1 * 25 ST
       195.00 SFr.
  • 22×68 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 232.00 SFr.
    232.00 SFr.
    1 * 25 ST
       232.00 SFr.
  • 22×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 188.00 SFr.
    188.00 SFr.
    1 * 25 ST
       188.00 SFr.
  • 25×100 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 225.00 SFr.
    225.00 SFr.
    1 * 25 ST
       225.00 SFr.
  • 26×60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 250.00 SFr.
    250.00 SFr.
    1 * 25 ST
       250.00 SFr.
  • 26×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 209.00 SFr.
    209.00 SFr.
    1 * 25 ST
       209.00 SFr.
  • 28×60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 261.00 SFr.
    261.00 SFr.
    1 * 25 ST
       261.00 SFr.
  • 30×70 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 212.00 SFr.
    212.00 SFr.
    1 * 25 ST
       212.00 SFr.
  • 30×77 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 209.00 SFr.
    209.00 SFr.
    1 * 25 ST
       209.00 SFr.
  • 30×100 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 226.00 SFr.
    226.00 SFr.
    1 * 25 ST
       226.00 SFr.
  • 33×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 217.00 SFr.
    217.00 SFr.
    1 * 25 ST
       217.00 SFr.
  • 33×94 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 228.00 SFr.
    228.00 SFr.
    1 * 25 ST
       228.00 SFr.
  • 35×90 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST
  • 35×150 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 267.00 SFr.
    267.00 SFr.
    1 * 25 ST
       267.00 SFr.
  • 40×150 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 290.00 SFr.
    290.00 SFr.
    1 * 25 ST
       290.00 SFr.
  • 43×123 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 326.00 SFr.
    326.00 SFr.
    1 * 25 ST
       326.00 SFr.
  • 44×230 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 342.00 SFr.
    342.00 SFr.
    1 * 25 ST
       342.00 SFr.
  • 53×145 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 373.00 SFr.
    373.00 SFr.
    1 * 25 ST
       373.00 SFr.