Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
Supplier:   PRAT DUMAS
Extraction thimbles
Supplier:  PRAT DUMAS
Certificates
Extraction thimbles
Extraction thimbles
16 Options Available Below
Product Details & Documents
Pure cotton cellulose
  • Neutral and free of fats and grease
  • Moulded in one piece
  • Very resistant to solvents
Applications include extraction of sludges, asphalt, foodstuffs and in the analysis and quality control of powders and dusts.
Product Family Options
Product Information
  • Int.Ø×H
    Availability
    Price
  • 80×200 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 730.00
    SFr. 730.00
    1 * 25 ST
       SFr. 730.00
  • 80×250 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 850.00
    SFr. 850.00
    1 * 25 ST
       SFr. 850.00
  • 58×170 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 489.00
    SFr. 489.00
    1 * 25 ST
       SFr. 489.00
  • 46×165 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 248.00
    SFr. 248.00
    1 * 25 ST
       SFr. 248.00
  • 41×150 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 201.00
    SFr. 201.00
    1 * 25 ST
       SFr. 201.00
  • 46×130 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 248.00
    SFr. 248.00
    1 * 25 ST
       SFr. 248.00
  • 34×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 130.00
    SFr. 130.00
    1 * 25 ST
       SFr. 130.00
  • 33×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 130.00
    SFr. 130.00
    1 * 25 ST
       SFr. 130.00
  • 30×60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 136.00
    SFr. 136.00
    1 * 25 ST
       SFr. 136.00
  • 27×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 121.00
    SFr. 121.00
    1 * 25 ST
       SFr. 121.00
  • 26×60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 115.00
    SFr. 115.00
    1 * 25 ST
       SFr. 115.00
  • 22×60 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 128.00
    SFr. 128.00
    1 * 25 ST
       SFr. 128.00
  • 70×240 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 558.00
    SFr. 558.00
    1 * 25 ST
       SFr. 558.00
  • 37×130 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 155.00
    SFr. 155.00
    1 * 25 ST
       SFr. 155.00
  • 30×100 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 144.00
    SFr. 144.00
    1 * 25 ST
       SFr. 144.00
  • 22×80 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 127.00
    SFr. 127.00
    1 * 25 ST
       SFr. 127.00