Orders LinkContactUsLinkComponent
VWR® Drum Keys
VWR® Drum Keys
Supplier:   VWR Collection
VWR® Drum Keys
Supplier:  VWR Collection
Certificates
VWR® Drum Keys
4 Options Available Below
Product Details & Documents
A selection of tools to assist in opening IBCs, 200 litre drums, plus bottles and other containers with DIN 32, 45, 54 and 80 caps.
Product Family Options
Product Information
  • Availability
    Price
  • 29548.053
    Drum keys, plastic, for tightening/opening caps for 20 or 25 L (DIN 61) plastic drums
    Drum keys, plastic, for tightening/opening caps for 20 or 25 L (DIN 61) plastic drums
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 12.80
    SFr. 12.80
    1 * 1 ST
       SFr. 12.80
  • 29548.064
    Drum keys, plastic, for tightening/opening caps for 10 L (DIN 51) plastic drums
    Drum keys, plastic, for tightening/opening caps for 10 L (DIN 51) plastic drums
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 14.00
    SFr. 14.00
    1 * 1 ST
       SFr. 14.00
  • VWRFCLEUNIVMETAL
    Drum key, metal, for seals and 2" and 3/4" closures on metal drums
    Drum key, metal, for seals and 2" and 3/4" closures on metal drums
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 87.10
    SFr. 87.10
    1 * 1 ST
       SFr. 87.10
  • 84544.001
    Drum key to open DIN 32/45/54/80 caps
    Drum key to open DIN 32/45/54/80 caps
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 26.50
    SFr. 26.50
    1 * 1 ST
       SFr. 26.50
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..