Order Entry
Switzerland
ContactUsLinkComponent
Laboratory trays, sterilising, Azlon®
Laboratory trays, sterilising, Azlon®
Catalog # 489-0001
Supplier:  SciLabware
undefined
Laboratory trays, sterilising, Azlon®
Catalog # 489-0001
Supplier:  SciLabware

Specifications

  • Capacity:
    10 l
  • Seq:
    02
  • W×D×H:
    290×390×110 mm
  • Pk:
    1

Specifications

About this item

PP, white.

  • Robust and deep to accommodate a wide range of instruments etc.
  • Autoclavable
  • Accessory lids are white polystyrene and are not autoclavable