Collagen I culture dishes, Corning® BioCoat™
: Corning
PS coated with rat tail collagen type I, sterile, non-pyrogenic.
- Manufactured in a highly controlled environment and rigorously tested to assure product consistency and performance
- Stable at room temperature when stored in dry conditions
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon and bone. It is an integral part of the framework that holds cells and tissues together and has been recognised as a useful matrix for improving cell culture. Used in vitro, collagen can exert effects on the adherence, morphology, growth, migration and differentiation of a variety of cell types.
- Catalog No:
- 734-0141
- 734-0280
- 734-0109
- 734-0202
- 734-0136
- 734-0275
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
Frequently Bought Together