Orders LinkContactUsLinkComponent
Dishes
Dishes
Supplier:   RSG
Dishes
Supplier:  RSG
Certificates
Dishes
9 Options Available Below
Product Details & Documents
Stainless steel, ¹⁸/₁₀.
  • With folded edge; fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, very good thermal resistance
Round multipurpose dishes for a wide range of uses.
Ordering information: Each dish set (Cat. No. 216-1249) comprises one of each 1, 2 and 3 litre dishes.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 400 ml
    45 mm
    155 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 22.70
    SFr. 22.70
    1 * 1 ST
       SFr. 22.70
  • 1 l
    90 mm
    175 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 24.90
    SFr. 24.90
    1 * 1 ST
       SFr. 24.90
  • 2 l
    115 mm
    217 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 29.90
    SFr. 29.90
    1 * 1 ST
       SFr. 29.90
  • 3 l
    120 mm
    245 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 36.00
    SFr. 36.00
    1 * 1 ST
       SFr. 36.00
  • 5 l
    135 mm
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 76.10
    SFr. 76.10
    1 * 1 ST
       SFr. 76.10
  • 6 l
    135 mm
    320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 86.50
    SFr. 86.50
    1 * 1 ST
       SFr. 86.50
  • 8 l
    160 mm
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 99.90
    SFr. 99.90
    1 * 1 ST
       SFr. 99.90
  • 11 l
    180 mm
    395 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 145.00
    SFr. 145.00
    1 * 1 ST
       SFr. 145.00
  • Set
    90/115/120 mm
    175/217/245 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 90.70
    SFr. 90.70
    1 * 1 ST
       SFr. 90.70