Orders LinkContactUsLinkComponent
Desiccators
Desiccators
Supplier:   KARTELL
Desiccators
Supplier:  KARTELL
Certificates
Desiccators
3 Options Available Below
Product Details & Documents
Transparent polycarbonate cover with polypropylene base. The specially grooved stopper ensures air is readmitted very slowly to avoid disturbing the contents.
  • Implosion proof
  • Will withstand vacuum of 740 mm Hg with loss no greater than 20 mm in 24 hours
  • Includes removable pan for holding drying agent
Delivery information: Supplied with vacuum release tap and O-ring.
Product Family Options
Product Information
  • Type
    Nominal size
    Usable height
    Ø ext.×H
    Availability
    Price
  • Desiccator
    200 DN
    175 mm
    235×230 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 205.00
    SFr. 205.00
    1 * 1 ST
       SFr. 205.00
  • Desiccator
    150 DN
    135 mm
    170×190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 196.00
    SFr. 196.00
    1 * 1 ST
       SFr. 196.00
  • Desiccator
    250 DN
    225 mm
    285×300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 243.00
    SFr. 243.00
    1 * 1 ST
       SFr. 243.00
Accessories
Product Information
  • Catalog #
    Type
    Availability
    Price
  • 467-2290
    Spare vacuum valve for desiccators
    Log in for availability
    1 * 1 ST - SFr. 27.20
Product Information
  • Catalog #
    Type
    Availability
    Price
  • 467-2215
    Rubber O-ring
    Log in for availability
    1 * 1 pezzi - SFr. 6.40
Product Information
  • Catalog #
    Type
    Availability
    Price
  • 467-2220
    Rubber O-ring
    Log in for availability
    1 * 1 ST - SFr. 8.70
Product Information
  • Catalog #
    Type
    Availability
    Price
  • 467-2225
    Rubber O-ring
    Log in for availability
    1 * 1 ST - SFr. 12.30