Orders LinkContactUsLinkComponent
Desiccators, Nalgene®
Desiccators, Nalgene®
Desiccators, Nalgene®
Desiccators, Nalgene®
Transparent PC lid and blue PP base with or without PTFE stopcock. Accepts 6,4 mm (¹/₄") int.Ø. vacuum tubing and can be used with any standard 230 mm desiccator plate, for example Cat. No. 467-3100. Desiccator 467-3110 should be used at room temperature only, 467-3112 is autoclavable.
  • Base is impervious to all solid desiccants, including acids and alkalies (do not use concentrated acids as desiccants)
  • No gasket or vacuum is needed to ensure cover is airtight, only requires grease
 : Supplied without plate, please order suitable plate separately.
  • Catalog No:
  • 467-3112
  • 467-3110
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Product Family Options
  • Type
    Nominal size
    Usable height
    Ø ext.×H
  • Without stopcock
    330 DN
    143 mm
    330×262 mm
    1 * 1 ST
    1 * 1 ST - SFr. 214.00
    SFr. 214.00
    1 * 1 ST
       SFr. 214.00
      Low Manufacturing Impact
    Specifications
    Cat. No.
    467-3112
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • With stopcock
    330 DN
    143 mm
    330×262 mm
    1 * 1 ST
    1 * 1 ST - SFr. 219.00
    SFr. 219.00
    1 * 1 ST
       SFr. 219.00
      Low Manufacturing Impact
    Specifications
    Cat. No.
    467-3110
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
Accessories
  •  
    Type
  • NALG71-5311-0250
    With recessed rim
    1 * 2 ST - SFr. 567.00