Orders LinkContactUsLinkComponent
Multi-purpose cylinders, DURAN®
Supplier:   DWK Life Sciences
Multi-purpose cylinders, DURAN®
Supplier:  DWK Life Sciences
Certificates
Multi-purpose cylinders, DURAN®
Multi-purpose cylinders, DURAN®
10 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with round base.
  • Very good chemical resistance and high resistance to temperature change
  • No graduations
  • Rough ground rim without pouring lip
Product Family Options
Product Information
  • Capacity
    Outer height
    Ext.Ø
    Availability
    Price
  • 180 ml
    200 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 20.60
    SFr. 20.60
    1 * 1 ST
       SFr. 20.60
  • 220 ml
    150 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 25.10
    SFr. 25.10
    1 * 1 ST
       SFr. 25.10
  • 280 ml
    300 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 24.70
    SFr. 24.70
    1 * 1 ST
       SFr. 24.70
  • 380 ml
    400 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 30.90
    SFr. 30.90
    1 * 1 ST
       SFr. 30.90
  • 420 ml
    200 mm
    60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 31.50
    SFr. 31.50
    1 * 1 ST
       SFr. 31.50
  • 450 ml
    300 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 29.80
    SFr. 29.80
    1 * 1 ST
       SFr. 29.80
  • 530 ml
    250 mm
    60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 34.90
    SFr. 34.90
    1 * 1 ST
       SFr. 34.90
  • 770 ml
    500 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 50.30
    SFr. 50.30
    1 * 1 ST
       SFr. 50.30
  • 1250 ml
    450 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 46.90
    SFr. 46.90
    1 * 1 ST
       SFr. 46.90
  • 1650 ml
    400 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 91.20
    SFr. 91.20
    1 * 1 ST
       SFr. 91.20