Seals for threaded connection caps, BOLA
Supplier: Bohlender
Certificates
About this item
Silicone with PTFE washers.
- Temperature resistant from −200 to +250 °C
- Very good chemical resistance
- Easy handling
For threaded connection caps with hole and GL thread.
Frequently Bought Together